2-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC302391-1G
PC302391-5G
APOSPC302391-1GEA
133
GBP
APOSPC302391-1G
APOSPC302391-5G
2-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%
2-Methyl-5-(trifluoromethoxy)phenylacetic acid
Formula:
C₁₀H₉F₃O₃ MW: 234,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399820 CAS Number: 1261573-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...