2,3-Dichloro-6-fluorobenzyl alcohol 98%

Supplier: Apollo Scientific

Synonyms: (2,3-Dichloro-6-fluorophenyl)methanol

PC302371-10G PC302371-5G
APOSPC302371-10GEA 317 GBP
APOSPC302371-10G APOSPC302371-5G
2,3-Dichloro-6-fluorobenzyl alcohol 98%
2,3-Dichloro-6-fluorobenzyl alcohol
Formula: C₇H₅Cl₂FO
MW: 195,018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115872
CAS Number: 886497-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR