5-Fluoro-2-(trifluoromethoxy)cinnamic acid 97%

Supplier: Apollo Scientific

PC302359-1G PC302359-5G PC302359-250MG
APOSPC302359-1GEA 348 GBP
APOSPC302359-1G APOSPC302359-5G APOSPC302359-250MG
5-Fluoro-2-(trifluoromethoxy)cinnamic acid 97%
5-Fluoro-2-(trifluoromethoxy)cinnamic acid
Formula: C₁₀H₆F₄O₃
MW: 250.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519340
CAS Number: 1092460-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR