2-Fluoro-4-methyl-DL-phenylalanine

Supplier: Apollo Scientific
Warning

PC302133-1G PC302133-5G
APOSPC302133-1GEA 106 GBP
APOSPC302133-1G APOSPC302133-5G
2-Fluoro-4-methyl-DL-phenylalanine
2-Fluoro-4-methyl-DL-phenylalanine
Formula: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116029
CAS Number: 1259965-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR