4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl alcohol 97%

Supplier: Apollo Scientific

PC302117-1G PC302117-5G
APOSPC302117-1GEA 130 GBP
APOSPC302117-1G APOSPC302117-5G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl alcohol 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzyl alcohol
Formula: C₁₀H₁₁F₃O₂
MW: 220,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795339
CAS Number: 1431329-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR