2,3-Difluoro-4-methylcinnamic acid 97%

Supplier: Apollo Scientific

PC302069-1G PC302069-5G
APOSPC302069-1GEA 154 GBP
APOSPC302069-1G APOSPC302069-5G
2,3-Difluoro-4-methylcinnamic acid 97%
2,3-Difluoro-4-methylcinnamic acid
Formula: C₁₀H₈F₂O₂
MW: 198,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03002806
CAS Number: 682804-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR