1-(3-Chloro-4-fluorophenyl)ethanol
Supplier: Apollo Scientific
PC300890-10G
PC300890-1G
PC300890-5G
APOSPC300890-10GEA
1020
GBP
APOSPC300890-10G
APOSPC300890-1G
APOSPC300890-5G
1-(3-Chloro-4-fluorophenyl)ethanol
1-(3-Chloro-4-fluorophenyl)ethanol
Formula:
C₈H₈ClFO MW: 174,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973766 CAS Number: 878572-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...