2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethanamine

Supplier: Apollo Scientific

PC300850-1G PC300850-250MG
APOSPC300850-1GEA 472 GBP
APOSPC300850-1G APOSPC300850-250MG
2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethanamine
2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethanamine
Formula: C₉H₁₀FN₃
MW: 179,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14546660
CAS Number: 1216295-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR