1-(4-Fluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Supplier: Apollo Scientific
|
Warning
|
PC28273-1G
PC28273-250MG
APOSPC28273-1GEA
108
GBP
APOSPC28273-1G
APOSPC28273-250MG
1-(4-Fluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
1-(4-Fluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Formula:
C₁₀H₈FN₃O MW: 205,188 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411212 CAS Number: 58905-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...