3-Chloro-5-(trifluoromethyl)picolinimidohydrazide

Supplier: Apollo Scientific
Warning

PC28272-1G PC28272-250MG
APOSPC28272-1GEA 426 GBP
APOSPC28272-1G APOSPC28272-250MG
3-Chloro-5-(trifluoromethyl)picolinimidohydrazide
3-Chloro-5-(trifluoromethyl)picolinimidohydrazide
Formula: C₇H₆ClF₃N₄
Storage Temperature: Ambient
MDL Number: MFCD31812111
CAS Number: 2379918-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR