4-Chloro-8-fluoro-6-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
|
Warning
|
PC28245-1G
PC28245-250MG
APOSPC28245-1GEA
318
GBP
APOSPC28245-1G
APOSPC28245-250MG
4-Chloro-8-fluoro-6-(trifluoromethyl)quinoline
4-Chloro-8-fluoro-6-(trifluoromethyl)quinoline
Formula:
C₁₀H₄ClF₄N MW: 249,592 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28010392 CAS Number: 1065093-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...