6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole

Supplier: Apollo Scientific

Synonyms: 6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole

PC28238-1G PC28238-5G
APOSPC28238-1GEA 73.5 GBP
APOSPC28238-1G APOSPC28238-5G
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole
Formula: C₁₁H₁₂BrFN₂
MW: 271,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660227
CAS Number: 1231930-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR