2-Bromo-4-fluorobenzyl fluoride 95%
Supplier: Apollo Scientific
|
Danger
|
PC27927-1G
PC27927-250MG
APOSPC27927-1GEA
551
GBP
APOSPC27927-1G
APOSPC27927-250MG
2-Bromo-4-fluorobenzyl fluoride 95%
2-Bromo-4-fluorobenzyl fluoride
Formula:
C₇H₅BrF₂ MW: 207,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22491732 CAS Number: 1823057-15-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...