(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid

Supplier: Apollo Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4-difluorobutanoic acid

PC27883-250MG
APOSPC27883-250MGEA 123 GBP
APOSPC27883-250MG
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid
Formula: C₁₉H₁₇F₂NO₄
MW: 361.339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796145
CAS Number: 467442-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR