2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4-fluorobutanoic acid
Supplier: Apollo Scientific
PC27862-100MG
PC27862-1G
PC27862-250MG
APOSPC27862-100MGEA
722
GBP
APOSPC27862-100MG
APOSPC27862-1G
APOSPC27862-250MG
2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4-fluorobutanoic acid
2-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-4-fluorobutanoic acid
Formula:
C₁₉H₁₈FNO₄ MW: 343,349 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31615633 CAS Number: 1690528-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...