3-Fluoro-4-(3,3,3-trifluoro-1-propyn-1-yl)benzoic acid 95%

Supplier: Apollo Scientific
Warning

PC27849-1G PC27849-250MG
APOSPC27849-1GEA 591 GBP
APOSPC27849-1G APOSPC27849-250MG
3-Fluoro-4-(3,3,3-trifluoro-1-propyn-1-yl)benzoic acid 95%
3-Fluoro-4-(3,3,3-trifluoro-1-propyn-1-yl)benzoic acid
Formula: C₁₀H₄F₄O₂
MW: 232,131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32264075
CAS Number: 2407339-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR