2-(Bromomethyl)perfluoropropan-2-ol 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(Bromomethyl)-2-hydroxyhexafluoropropane, 2-(Bromomethyl)hexafluoropropan-2-ol, 1,1-Bis(trifluoromethyl)-2-bromoethan-1-ol
PC2652-1G
PC2652-5G
PC2652-250MG
APOSPC2652-1GEA
195
GBP
APOSPC2652-1G
APOSPC2652-5G
APOSPC2652-250MG
2-(Bromomethyl)perfluoropropan-2-ol 97%
2-(Bromomethyl)perfluoropropan-2-ol
Formula:
C₄H₃BrF₆O MW: 260,96 g/mol Boiling Pt: 101…103 °C Density: 1,9 g/cm³ (25 °C) Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD08460483 CAS Number: 503169-76-4 UN: 2924 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...