3-(4-Fluorophenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione

Supplier: Apollo Scientific

PC210053-1G PC210053-500MG
APOSPC210053-1GEA 486 GBP
APOSPC210053-1G APOSPC210053-500MG
3-(4-Fluorophenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
3-(4-Fluorophenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
Formula: C₁₄H₁₅FN₂S
MW: 262,346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14735652
CAS Number: 899926-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR