N-(3,4-Difluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
Supplier: Apollo Scientific
PC210049-1G
PC210049-500MG
APOSPC210049-1GEA
486
GBP
APOSPC210049-1G
APOSPC210049-500MG
N-(3,4-Difluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
N-(3,4-Difluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-sulphonamide
Formula:
C₁₂H₈F₂N₄O₂S MW: 310,279 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18804117 CAS Number: 1291486-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...