(3-Bromo-2-fluorophenyl)methanamine 97%

Supplier: Apollo Scientific

PC201217-100MG PC201217-1G PC201217-250MG
APOSPC201217-100MGEA 112 GBP
APOSPC201217-100MG APOSPC201217-1G APOSPC201217-250MG
(3-Bromo-2-fluorophenyl)methanamine 97%
(3-Bromo-2-fluorophenyl)methanamine
Formula: C₇H₇BrFN
MW: 204,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194194
CAS Number: 261723-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR