3-Fluoro-N-methoxy-N-methylbenzamide 95+%
Supplier: Apollo Scientific
PC201203-1G
PC201203-5G
APOSPC201203-1GEA
52.5
GBP
APOSPC201203-1G
APOSPC201203-5G
3-Fluoro-N-methoxy-N-methylbenzamide 95+%
3-Fluoro-N-methoxy-N-methylbenzamide
Formula:
C₉H₁₀FNO₂ MW: 183,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02684305 CAS Number: 226260-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...