(S)-2-Amino-2-(4-fluorophenyl)ethanol 97%

Supplier: Apollo Scientific

PC201202-1G PC201202-250MG PC201202-5G
APOSPC201202-1GEA 257 GBP
APOSPC201202-1G APOSPC201202-250MG APOSPC201202-5G
(S)-2-Amino-2-(4-fluorophenyl)ethanol 97%
(S)-2-Amino-2-(4-fluorophenyl)ethanol
Formula: C₈H₁₀FNO
MW: 155,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09253714
CAS Number: 224434-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR