5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine 95+%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-2-(2,2,2-trifluoro ethoxy)pyridine

PC201108-1G PC201108-250MG
APOSPC201108-1GEA 80 GBP
APOSPC201108-1G APOSPC201108-250MG
5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine 95+%
5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine
Formula: C₇H₅BrF₃NO
MW: 256,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374892
CAS Number: 126728-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR