5-(2-Fluorophenyl)-1H-pyrrole-3-carbonitrile 95+%
Supplier: Apollo Scientific
PC201087-1G
PC201087-250MG
APOSPC201087-1GEA
64.3
GBP
APOSPC201087-1G
APOSPC201087-250MG
5-(2-Fluorophenyl)-1H-pyrrole-3-carbonitrile 95+%
5-(2-Fluorophenyl)-1H-pyrrole-3-carbonitrile
Formula:
C₁₁H₇FN₂ MW: 186,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24619247 CAS Number: 1240948-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...