(2-Bromo-3-fluoropyridin-4-yl)methanol 95+%

Supplier: Apollo Scientific

PC201081-1G PC201081-250MG
APOSPC201081-1GEA 304 GBP
APOSPC201081-1G APOSPC201081-250MG
(2-Bromo-3-fluoropyridin-4-yl)methanol 95+%
(2-Bromo-3-fluoropyridin-4-yl)methanol
Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606993
CAS Number: 1227601-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR