Methyl 3-bromo-5-fluoroisonicotinate 97%

Supplier: Apollo Scientific

Synonyms: Methyl 3-Bromo-5-fluoro-4-pyridinecarboxylate

PC201068-100MG PC201068-250MG
APOSPC201068-100MGEA 98.3 GBP
APOSPC201068-100MG APOSPC201068-250MG
Methyl 3-bromo-5-fluoroisonicotinate 97%
Methyl 3-bromo-5-fluoroisonicotinate
Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698118
CAS Number: 1214325-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR