Potassium trifluoro({4-[methoxy(methyl)carbamoyl]phenyl})boranuide

Supplier: Apollo Scientific

PC200160-1G PC200160-500MG PC200160-5G
APOSPC200160-1GEA 106 GBP
APOSPC200160-1G APOSPC200160-500MG APOSPC200160-5G
Potassium trifluoro({4-[methoxy(methyl)carbamoyl]phenyl})boranuide
Potassium trifluoro({4-[methoxy(methyl)carbamoyl]phenyl})boranuide
Formula: C₉H₁₀BF₃NO₂·K
MW: 271,086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294523
CAS Number: 1644635-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR