Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate 95+%

Supplier: Apollo Scientific

PC1843-100MG PC1843-1G PC1843-250MG
APOSPC1843-100MGEA 78.7 GBP
APOSPC1843-100MG APOSPC1843-1G APOSPC1843-250MG
Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate 95+%
Ethyl 3-(2,5-difluorophenyl)-3-oxopropanoate
Formula: C₁₁H₁₀F₂O₃
MW: 228,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783808
CAS Number: 887267-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR