3-Fluoro-4-(trifluoromethoxy)phenylacetylene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Ethynyl-2-fluoro-1-(trifluoromethoxy)benzene, 4-Ethynyl-α,α,α,2-tetrafluoroanisole
PC1161-5G
PC1161-1G
APOSPC1161-5GEA
159
GBP
APOSPC1161-5G
APOSPC1161-1G
3-Fluoro-4-(trifluoromethoxy)phenylacetylene 98%
3-Fluoro-4-(trifluoromethoxy)phenylacetylene
Formula:
C₉H₄F₄O MW: 204,121 g/mol Density: 1,279 g/cm³ (25 °C) Flash Pt: 40 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08458006 CAS Number: 912617-68-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...