3-Amino-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Supplier: Apollo Scientific
Warning

PC110016-1G PC110016-250MG
APOSPC110016-1GEA 236 GBP
APOSPC110016-1G APOSPC110016-250MG
3-Amino-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Amino-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formula: C₁₀H₆F₃NO₂S
MW: 261,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29169135
CAS Number: 2379918-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR