2'-Amino-4'-chloro-2,2,2-trifluoroacetophenone

Supplier: Apollo Scientific
Warning

PC10723-250MG PC10723-100MG
APOSPC10723-250MGEA 166 GBP
APOSPC10723-250MG APOSPC10723-100MG
2'-Amino-4'-chloro-2,2,2-trifluoroacetophenone
2'-Amino-4'-chloro-2,2,2-trifluoroacetophenone
Formula: C₈H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD21781429
CAS Number: 1448858-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR