3-Chloro-4-(trifluoromethoxy)benzamide 98%

Supplier: Apollo Scientific

PC0176-5G PC0176-1G
APOSPC0176-5GEA 178 GBP
APOSPC0176-5G APOSPC0176-1G
3-Chloro-4-(trifluoromethoxy)benzamide 98%
3-Chloro-4-(trifluoromethoxy)benzamide
Formula: C₈H₅ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01631466
CAS Number: 40251-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR