1,1,1-Trifluorohexane 97%

Supplier: Apollo Scientific
Danger

PC01657-1G PC01657-5G
APOSPC01657-1GEA 65.6 GBP
APOSPC01657-1G APOSPC01657-5G
1,1,1-Trifluorohexane 97%
1,1,1-Trifluorohexane
Formula: C₆H₁₁F₃
MW: 140,147 g/mol
Boiling Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD16606064
CAS Number: 17337-12-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR