[2-(Fur-2-yl)phenyl]methylamine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Fur-2-yl)benzylamine, 2-[2-(Aminomethyl)phenyl]furan

OR9675-1G OR9675-250MG
APOSOR9675-1GEA 237 GBP
APOSOR9675-1G APOSOR9675-250MG
[2-(Fur-2-yl)phenyl]methylamine 97%
[2-(Fur-2-yl)phenyl]methylamine
Formula: C₁₁H₁₁NO
MW: 173,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213664
CAS Number: 771573-25-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR