4-(1,3,4-Oxadiazol-2-yl)phenol 98%

Supplier: Apollo Scientific

OR965587-10G OR965587-1G OR965587-5G
APOSOR965587-10GEA 780 GBP
APOSOR965587-10G APOSOR965587-1G APOSOR965587-5G
4-(1,3,4-Oxadiazol-2-yl)phenol 98%
4-(1,3,4-Oxadiazol-2-yl)phenol
Formula: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221056
CAS Number: 5378-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR