2-(Methylsulphonamido)benzaldehyde 97%

Supplier: Apollo Scientific

OR965391-1G OR965391-5G
APOSOR965391-1GEA 157 GBP
APOSOR965391-1G APOSOR965391-5G
2-(Methylsulphonamido)benzaldehyde 97%
2-(Methylsulphonamido)benzaldehyde
Formula: C₈H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD11052341
CAS Number: 94532-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR