3,4-Dihydro-1H-2-benzopyran-1-carboxylic acid 95%

Supplier: Apollo Scientific

OR964437-1G OR964437-250MG
APOSOR964437-1GEA 320 GBP
APOSOR964437-1G APOSOR964437-250MG
3,4-Dihydro-1H-2-benzopyran-1-carboxylic acid 95%
3,4-Dihydro-1H-2-benzopyran-1-carboxylic acid
Formula: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00457636
CAS Number: 13328-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR