4-(Boc-amino)benzyl alcohol 97%

Supplier: Apollo Scientific

Synonyms: tert-Butyl (4-(hydroxymethyl)phenyl)carbamate

OR962629-1G OR962629-5G
APOSOR962629-1GEA 48.6 GBP
APOSOR962629-1G APOSOR962629-5G
4-(Boc-amino)benzyl alcohol 97%
4-(Boc-amino)benzyl alcohol
Formula: C₁₂H₁₇NO₃
MW: 223,268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974271
CAS Number: 144072-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR