2-(Cyclohexyloxy)acetic acid 98%

Supplier: Apollo Scientific

OR962599-1G OR962599-5G
APOSOR962599-1GEA 171 GBP
APOSOR962599-1G APOSOR962599-5G
2-(Cyclohexyloxy)acetic acid 98%
2-(Cyclohexyloxy)acetic acid
Formula: C₈H₁₄O₃
MW: 158,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001653
CAS Number: 71995-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR