1-Phenyl-1H-1,2,4-triazole 95%
Supplier: Apollo Scientific
OR962488-1G
OR962488-250MG
APOSOR962488-1GEA
199
GBP
APOSOR962488-1G
APOSOR962488-250MG
1-Phenyl-1H-1,2,4-triazole 95%
1-Phenyl-1H-1,2,4-triazole
Formula:
C₈H₇N₃ MW: 145,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01076193 CAS Number: 13423-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...