4-Methoxy-2,3-dihydro-1H-indole 95%

Supplier: Apollo Scientific

Synonyms: 4-Methoxyindoline

OR962408-100MG OR962408-1G OR962408-250MG
APOSOR962408-100MGEA 68.3 GBP
APOSOR962408-100MG APOSOR962408-1G APOSOR962408-250MG
4-Methoxy-2,3-dihydro-1H-indole 95%
4-Methoxy-2,3-dihydro-1H-indole
Formula: C₉H₁₁NO
MW: 149,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371625
CAS Number: 7555-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR