2-(2-Chloro-phenyl)-propionitrile 95%

Supplier: Apollo Scientific

OR962319-1G OR962319-250MG
APOSOR962319-1GEA 617 GBP
APOSOR962319-1G APOSOR962319-250MG
2-(2-Chloro-phenyl)-propionitrile 95%
2-(2-Chloro-phenyl)-propionitrile
Formula: C₉H₈ClN
MW: 165,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687167
CAS Number: 75920-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR