Thiazolidine hydrochloride 95%
Supplier: Apollo Scientific
OR961766-25G
OR961766-5G
APOSOR961766-25GEA
420
GBP
APOSOR961766-25G
APOSOR961766-5G
Thiazolidine hydrochloride 95%
Thiazolidine hydrochloride
Formula:
C₃H₇NS·ClH MW: 125,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20483376 CAS Number: 14446-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...