H-Ala-oall hydrochloride 98%
Supplier: Apollo Scientific
OR961118-10G
OR961118-1G
OR961118-25G
APOSOR961118-10GEA
296
GBP
APOSOR961118-10G
APOSOR961118-1G
APOSOR961118-25G
H-Ala-oall hydrochloride 98%
H-Ala-oall hydrochloride
Formula:
C₆H₁₁NO₂·ClH MW: 165.618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01074682 CAS Number: 203799-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...