7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 97%

Supplier: Apollo Scientific

OR960206-10G OR960206-1G OR960206-5G
APOSOR960206-10GEA 681 GBP
APOSOR960206-10G APOSOR960206-1G APOSOR960206-5G
7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 97%
7-Chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
Formula: C₉H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD26381971
CAS Number: 1506636-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR