1-(Cyclopropylmethyl)pyrazole 97%
Supplier: Apollo Scientific
OR959881-10G
OR959881-1G
OR959881-5G
APOSOR959881-10GEA
486
GBP
APOSOR959881-10G
APOSOR959881-1G
APOSOR959881-5G
1-(Cyclopropylmethyl)pyrazole 97%
1-(Cyclopropylmethyl)pyrazole
Formula:
C₇H₁₀N₂ MW: 122,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18889145 CAS Number: 1344382-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...