1-Propylpyrazole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Propyl-1H-pyrazole

OR959874-100G OR959874-10G OR959874-25G
APOSOR959874-100GEA 611 GBP
APOSOR959874-100G APOSOR959874-10G APOSOR959874-25G
1-Propylpyrazole 98%
1-Propylpyrazole
Formula: C₆H₁₀N₂
MW: 110,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08696396
CAS Number: 32500-67-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR