tert-Butyl pyridine-2-carboxylate 98%
Supplier: Apollo Scientific
OR959821-1G
OR959821-5G
APOSOR959821-1GEA
197
GBP
APOSOR959821-1G
APOSOR959821-5G
tert-Butyl pyridine-2-carboxylate 98%
tert-Butyl pyridine-2-carboxylate
Formula:
C₁₀H₁₃NO₂ MW: 179,216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19600064 CAS Number: 79233-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...