methyl 5-formylpyridine-2-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 5-Formylpyridine-2-carboxylate

OR959539-100MG OR959539-1G OR959539-250MG
APOSOR959539-100MGEA 53.8 GBP
APOSOR959539-100MG APOSOR959539-1G APOSOR959539-250MG
methyl 5-formylpyridine-2-carboxylate 95%
methyl 5-formylpyridine-2-carboxylate
Formula: C₈H₇NO₃
MW: 165,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100669
CAS Number: 55876-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR