2-(Pyrrolidin-1-yl)pyrimidin-4-amine 98%

Supplier: Apollo Scientific

OR959528-1G OR959528-5G
APOSOR959528-1GEA 243 GBP
APOSOR959528-1G APOSOR959528-5G
2-(Pyrrolidin-1-yl)pyrimidin-4-amine 98%
2-(Pyrrolidin-1-yl)pyrimidin-4-amine
Formula: C₈H₁₂N₄
Storage Temperature: Ambient
MDL Number: MFCD16103673
CAS Number: 33851-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR